Learn More
Andwin Scientific Pipetus pipet filler with color display, 0.1 mL to 200
SDP
Catalog No. NC0359337
Encompass_Preferred
The pipetus pipet filler features a color display that shows the battery level, speed level, and pipetting mode. This product provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL.
Catalog No. NC0359337 Supplier Andwin Scientific Supplier No. 9907200
Add to Cart
Add to Cart
The pipetus pipet filler features a color display that shows the battery level, speed level, and pipetting mode. This product provides highly accurate pipetting. It can also be used for measuring, volumetric and Pasteur pipettes, from 0.1 mL to 200 mL.