Learn More
Sigma Aldrich Fine Chemicals Biosciences HirschmannR pipetusR recha1EA
Catalog No. 502905861
Encompass
A high tech light weight (200Ag) ergonomically designed pipette pump with complete control at a single glance - the colored LCD display is the control center for all features of the new pipettus-akkuRechargeable battery up to 50Ahour use with charge indicatorUse with volumetric serological and plastic pipettes volumes 0.1 to 200AmLDisplay of selected pipetting mode EX or blow-outIndication of the selected pipetting speedBuzzer signal sounds when battery capacity reaches below 10%Adjustable suction speed depending upon volume and viscosity of mediumOne button operationNo trailing wiresfine dosing valveSilicone adapter with exchangeable internal filter holds all types of pipettes firm and tightCharges automatically just by inserting into stand
Catalog No. 50-290-5861 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3149511EA
A high tech light weight (200Ag) ergonomically designed pipette pump with complete control at a single glance - the colored LCD display is the control center for all features of the new pipettus-akkuRechargeable battery up to 50Ahour use with charge indicatorUse with volumetric serological and plastic pipettes volumes 0.1 to 200AmLDisplay of selected pipetting mode EX or blow-outIndication of the selected pipetting speedBuzzer signal sounds when battery capacity reaches below 10%Adjustable suction speed depending upon volume and viscosity of mediumOne button operationNo trailing wiresfine dosing valveSilicone adapter with exchangeable internal filter holds all types of pipettes firm and tightCharges automatically just by inserting into stand