Learn More
Chemglass Life Sciences Filter Funnel, Hirsh, 50mm Diameter Funnel Top, 20mm Coarse Frit
SDP
Catalog No. CG140802
Encompass_Preferred
Filter Funnel, Hirsh, 50mm Diameter Funnel Top, 20mm Coarse Frit. Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper. The 60º angle on the funnel permits removal of precipitates.
Catalog No. CG140802 Supplier Chemglass Life Sciences Supplier No. CG140802
Filter Funnel, Hirsh, 50mm Diameter Funnel Top, 20mm Coarse Frit. Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper. The 60º angle on the funnel permits removal of precipitates.