Learn More
Bachem Hirudin (54-65) (desulfated) Trifluoroacetate
SDP
Supplier:  Bachem 4014556.0001
Encompass_Preferred
Hirudin (54-65) (desulfated) Trifluoroacetate, H-Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr-Leu-Gln-OH, 113274-56-9, C66H93N13O25, Mr 1468.54, 1mg.
Catalog No. 50-383-363