Learn More
Sigma Aldrich Fine Chemicals Biosciences Hirudin from leeches lyoph
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H7016100UN
Encompass
Hirudin is a 7 kDa acidic protein containing 65 amino acid residues that has an isoelectric point of 3.5-4.0. It is not glycosylated and lacks tryptophan arginine and methionine residues. It is chemically stable between pH 2-12 and at temperatures up to 80C.
Catalog No. 50-290-2109