Learn More
Aniara Diagnostica LLC Recombinant Hirudin, 16,000 ATU per vial, lyophilized, for research use only
Supplier: Aniara Diagnostica LLC ARE120A
Recombinant Hirudin (ARE120A) is an effective thrombin inhibitor from Aniara Diagnostica LLC, specifically designed for in vitro laboratory research. This protein binds to thrombin at a 1:1 ratio, efficiently inhibiting both soluble and clot-bound thrombin, making it a crucial component for scientific studies focused on anticoagulation. Supplied in a lyophilized form for stability, this product is strictly for research use and not intended for any diagnostic applications.
- 16,000 ATU activity per vial
- Approximate reconstituted activity of 16,000 ATU/ml
- Lyophilized presentation for stability
- Requires reconstitution with 1 ml distilled water
- Storage: 2 days at room temperature; 7 days at 2-8°C; frozen at -20°C or below
- Contains glycine, Hepes, sodium chloride as excipients
- Amino acid sequence comprises 65 residues
- Protein size is around 7 kDa
- Use in in vitro applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.