Learn More
Cayman Chemical HIrudIn54-65non-sulfated 1mg
SDP
Catalog No. 502946707
Encompass_Preferred
A desulfated peptide fragment of hirudin; inhibits isolated bovine thrombin-induced fibrin clot formation (IC50 = 3.7 µM)
Catalog No. 50-294-6707 Supplier Cayman Chemical Supplier No. 337161 MG
A desulfated peptide fragment of hirudin; inhibits isolated bovine thrombin-induced fibrin clot formation (IC50 = 3.7 µM)