Learn More
Zymo Research Corporation His-Affinity Gel, 14 ml
SDP
Catalog No. 501251653
Encompass_Preferred
Nickel affinity gel used for the purification of histidine-tagged proteins-6% beaded agarose. 50% suspension in 30% ethanol. Protein binding capacity is 15mg/ml
Catalog No. 50-125-1653 Supplier Zymo Research Corporation Supplier No. P20032
Nickel affinity gel used for the purification of histidine-tagged proteins-6% beaded agarose. 50% suspension in 30% ethanol. Protein binding capacity is 15mg/ml