Learn More
Sigma Aldrich Fine Chemicals Biosciences His Buffer Kit Cytiva 11-0
Catalog No. 502901700
Encompass
His Buffer Kit contains carefully prepared buffer concentrates for binding washing and elution of histidine-tagged proteins according to recommended protocols. Moreover method for optimization of imidazole concentration in the binding buffer is described.
Catalog No. 50-290-1700 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE11003400
His Buffer Kit contains carefully prepared buffer concentrates for binding washing and elution of histidine-tagged proteins according to recommended protocols. Moreover method for optimization of imidazole concentration in the binding buffer is described.