Learn More
Medchemexpress LLC His-D-beta-Nal-Ala-Trp-D-Phe-Lys-NH2 TFA | 99.6% | 998.06 | 1 MG
SDP
Catalog No. 5000399723
Encompass_Preferred
His-D-beta-Nal-Ala-Trp-D-Phe-Lys-NH2 TFA is identified as a growth hormone releasing peptide and a metabolite of GHRP-1. GHRP-1 (Ala-His-D-beta Nal-Ala-Trp-D-Phe-Lys-NH2) promotes the release of growth hormone (GH) and increases [Ca2+]i levels in static monolayer cells of rat pituitary gland, without affecting cAMP levels.
  • Growth hormone releasing peptide
  • Metabolite of GHRP-1
  • Promotes the release of growth hormone (GH)
  • Increases [Ca2+]i levels in static monolayer cells of rat pituitary gland
Catalog No. 50-003-99723 Supplier Medchemexpress LLC Supplier No. HYP5189A1MG
His-D-beta-Nal-Ala-Trp-D-Phe-Lys-NH2 TFA is identified as a growth hormone releasing peptide and a metabolite of GHRP-1. GHRP-1 (Ala-His-D-beta Nal-Ala-Trp-D-Phe-Lys-NH2) promotes the release of growth hormone (GH) and increases [Ca2+]i levels in static monolayer cells of rat pituitary gland, without affecting cAMP levels.
  • Growth hormone releasing peptide
  • Metabolite of GHRP-1
  • Promotes the release of growth hormone (GH)
  • Increases [Ca2+]i levels in static monolayer cells of rat pituitary gland