Learn More
Zymo Research Corporation His-Elution Buffer, 25 ml
SDP
Catalog No. 501251656
Encompass_Preferred
His-Elution Buffer (25 ml). For use with His-Spin Protein Miniprep™ Kit for the purification of His-tagged proteins from cell free extracts.
Catalog No. 50-125-1656 Supplier Zymo Research Corporation Supplier No. P20035
His-Elution Buffer (25 ml). For use with His-Spin Protein Miniprep™ Kit for the purification of His-tagged proteins from cell free extracts.