Learn More
Epigentek Inc Histone H3R8 Dimethyl Symmetric (H3R8me2s) Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A3706100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human Asymmetric DiMethyl-Histone H3-R26. Tested Applications: WB, IHC, IF, IP. Reactivity: Human, Mouse, Rat, Broad Range. Alternative Names: H3R26me2a, HIST1H3J, H3/j , H3FJ , Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h , Histone H3/I, Histone H3/j , Histone H3/k, Histone H3/l, HIST3H3, H3 Arginine 26 me2a.
Catalog No. 50-109-5394