Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC His-Pro hydrochloride | 2415727-78-3 | 99.8% | 288.73 | 100 MG
SDP

Catalog No. 5000439245
Encompass_Preferred

His-Pro (Histidyl-proline) hydrochloride is a dipeptide consisting of histidyl and proline. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Sealed storage, away from moisture and light. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility (In Vitro): DMSO: 125 mg/mL (432.93 mM)

Catalog No. 50-004-39245 Supplier Medchemexpress LLC Supplier No. HY111659A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

His-Pro (Histidyl-proline) hydrochloride is a dipeptide consisting of histidyl and proline. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Sealed storage, away from moisture and light. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Solubility (In Vitro): DMSO: 125 mg/mL (432.93 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.