Learn More
Santa Cruz Biotechnology HIS PROB H3-HRP
Supplier:  Santa Cruz Biotechnology SC8036 HRP
Encompass
His-probe (H-3), Mouse monoclonal IgG1, 200ug/mL, Raised against a his tagged recombinant Protein, Recommended for detection of fusion Proteins encoded by polyhistidine expression vectors by WB and IP
Catalog No. NC9189892