Learn More
Sigma Aldrich Fine Chemicals Biosciences HIS SelectR HF Nckel Afi500ML
Catalog No. 502908980
Encompass
HIS-Select High Flow Nickel Affinity Gel is an immobilized metal-ion affinity chromatography (IMAC) gel that is designed to specifically bind His-tag proteins (histidine containing proteins) in chromatographic systems with pressures up to 200 psi and a maximum linear flow rate of 3000 cm/hr.
Catalog No. 50-290-8980 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H0537500ML
Add to Cart
Add to Cart
Request a Quote
HIS-Select High Flow Nickel Affinity Gel is an immobilized metal-ion affinity chromatography (IMAC) gel that is designed to specifically bind His-tag proteins (histidine containing proteins) in chromatographic systems with pressures up to 200 psi and a maximum linear flow rate of 3000 cm/hr.