Learn More
Sigma Aldrich Fine Chemicals Biosciences HIS SelectR Nickel Affinit
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P661125ML
Encompass
HIS-Select(R) Nickel Affinity Gel is durable and can capture the recombinant proteins with histidine tags at a high flow rate. Recombinant proteins with histidine tags are bound using either native or denaturing conditions.
Catalog No. 50-290-4822