Learn More
Zymo Research Corporation His-Spin Protein Miniprep™ Kit (50 Preps)
SDP
Catalog No. NC9227397
Encompass_Preferred
The simplified procedure is based on our innovative protein purification chemistry and custom designed fast spin columns. Up to 1 mg of His tagged protein can be purified in 5 minutes and eluted in as little as 100 ul of His-Elution Buffer. The purifiedprotein can be used directly for enzymatic assays, protein biochemical analyses, SDS-PAGE and other applications. The product has been optimized for maximal protein purity: a single protein band is visible by Coomassie blue staining on SDS-PAGE gel. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.
Catalog No. NC9227397 Supplier Zymo Research Corporation Supplier No. P2002
The simplified procedure is based on our innovative protein purification chemistry and custom designed fast spin columns. Up to 1 mg of His tagged protein can be purified in 5 minutes and eluted in as little as 100 ul of His-Elution Buffer. The purifiedprotein can be used directly for enzymatic assays, protein biochemical analyses, SDS-PAGE and other applications. The product has been optimized for maximal protein purity: a single protein band is visible by Coomassie blue staining on SDS-PAGE gel. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.