Learn More
Zymo Research Corporation His-Spin Protein Miniprep™ Kit (10 Preps)
SDP
Catalog No. NC0876634
Encompass_Preferred
The simplified procedure is based on ourinnovative protein purification chemistry and customdesigned fast spin columns. Up to 1 mg of His taggedprotein can be purified in 5 minutes and eluted in aslittle as 100 ul of His-Elution Buffer. The purifiedprotein can be used directly for enzymatic assays,protein biochemical analyses, SDS-PAGE and otherapplications. The product has been optimized formaximal protein purity: a single protein band is visibleby Coomassie blue staining on SDS-PAGE gel. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.
Catalog No. NC0876634 Supplier Zymo Research Corporation Supplier No. P2001
The simplified procedure is based on ourinnovative protein purification chemistry and customdesigned fast spin columns. Up to 1 mg of His taggedprotein can be purified in 5 minutes and eluted in aslittle as 100 ul of His-Elution Buffer. The purifiedprotein can be used directly for enzymatic assays,protein biochemical analyses, SDS-PAGE and otherapplications. The product has been optimized formaximal protein purity: a single protein band is visibleby Coomassie blue staining on SDS-PAGE gel. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.