Learn More
Sigma Aldrich Fine Chemicals Biosciences His SpintrapTM Cytiva 28 4
Catalog No. 502904469
Encompass
His SpinTrap(TM) is a prepacked single-use spin column for purifying histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC)* See the section for licensing information in our terms and conditions.Purification of unclarified samples on His SpinTrap(TM) columns minimizes loss of target protein caused by manual operations such as sample precentrifugation transfer to centrifugation tubes and collecting supernatant. In addition loading unclarified sample directly to His SpinTrap(TM) columns reduces sample preparation time which minimizes degradation and oxidation of sensitive target proteins.His SpinTrap(TM) columns are used together with a standard microcentrifuge and one purification run takes approximately 10 min. for optimal performance use His SpinTrap(TM) together with buffers prepared using His Buffer Kit.The columns are prepacked with Ni Sepharose(R) High Performance which is precharged with nickel ions for capture of histidine-tagged proteins. Leaka
Catalog No. 50-290-4469 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28401353
His SpinTrap(TM) is a prepacked single-use spin column for purifying histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC)* See the section for licensing information in our terms and conditions.Purification of unclarified samples on His SpinTrap(TM) columns minimizes loss of target protein caused by manual operations such as sample precentrifugation transfer to centrifugation tubes and collecting supernatant. In addition loading unclarified sample directly to His SpinTrap(TM) columns reduces sample preparation time which minimizes degradation and oxidation of sensitive target proteins.His SpinTrap(TM) columns are used together with a standard microcentrifuge and one purification run takes approximately 10 min. for optimal performance use His SpinTrap(TM) together with buffers prepared using His Buffer Kit.The columns are prepacked with Ni Sepharose(R) High Performance which is precharged with nickel ions for capture of histidine-tagged proteins. Leaka