Learn More
Genscript Corporation His Tag ELISA Detection Kit
Supplier:  Genscript Corporation L00436
Encompass
GenScript His Tag ELISA Detection Kit, a 1.5 hour competition ELISA, is developed for rapid and high throughput detection of His-tagged proteins. There are several potential usages:  Quickly identify the presence of His-tagged proteins in samples.  Optimize protein expression by monitoring the His-tagged proteins level.  High throughput screening of stable cell lines expressing His-tagged proteins.
Catalog No. NC0958221