Learn More
Abcam His-Tag Protein Purification Column (Pre-packed, 5 x 1ml) - AminTrap, 5UNIT
Supplier:  Abcam AB2705285UNIT
Encompass_Preferred
His-Tag Protein Purification Column (Pre-packed, 5 x 1ml) - AminTrap (ab270528) is a simple and easy ready to use chromatography medium for purifying His-tagged proteins that have been expressed in series of expression vectors, such as E.coli., yeast, insect cells and mammalian cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2058282