Learn More
Bioworld His-Tagged protein purification, Ni Separopore Kit
Supplier:  Bioworld 208300171
Encompass
bio-World's His-Tagged protein purification Kit consists of Ni-Chelated Separopore (4% beaded agarose), animminodiacetate (IDA)-immobilized metal affinity chromatography (IMAC) matrix that separates proteins via their differences in affinity to nickel metal ions. Kit includes Ni matrix and all the buffers needed for convenient protein purificatin.Ideal for purification of recombinant proteins containing (His)6 fusion tag.Kit Components:• Ni-Chelated Separopore, 3X 1ml columns• Binding/Wash buffer• Elution buffer
Catalog No. 50-198-439