Learn More
Enzo Life Sciences His(6) Recombinant monoclonal antibody (Rat IgG2b kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5360200
Encompass_Preferred
Clone: 3D5. Isotype: IgG2bκ. Immunogen: Purified his tagged Fab fragment. Specificity: Recognizes carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags. Applications: ELISA, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3062