Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScale 16/40
Catalog No. 502909130
Encompass
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter simplifying adjustments as well as disassembly and cleaning. The measurement scale on the column makes it easier to predict and reach a predefined packing height thereby increasing process control as well as reproducibility. Although optimized for high-pressure media HiScale columns are fully compatible with all applications (and packing methods) currently applicable to XK columns.
Catalog No. 50-290-9130 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28964424
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter simplifying adjustments as well as disassembly and cleaning. The measurement scale on the column makes it easier to predict and reach a predefined packing height thereby increasing process control as well as reproducibility. Although optimized for high-pressure media HiScale columns are fully compatible with all applications (and packing methods) currently applicable to XK columns.