Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScreen Capto Adhere ImpR
Catalog No. 502904075
Encompass
The multimodal functionality of Capto adhere ImpRes gives a different selectivity compared with traditional ion exchange columns and it binds proteins at high or low ionic strength.Contaminants such as DNA host cell proteins (HCP) leached protein A aggregates and viruses are efficiently removed making Capto adhere ImpRes an excellent choice for purification of MAbs and other biomolecules with high aggregate content.Capto adhere ImpRes is designed to allow effective high-reSOLUTIONon polishing of MAb in the second or third step of a purification scheme downstream of the protein A capture step.
Catalog No. 50-290-4075 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17371520
The multimodal functionality of Capto adhere ImpRes gives a different selectivity compared with traditional ion exchange columns and it binds proteins at high or low ionic strength.Contaminants such as DNA host cell proteins (HCP) leached protein A aggregates and viruses are efficiently removed making Capto adhere ImpRes an excellent choice for purification of MAbs and other biomolecules with high aggregate content.Capto adhere ImpRes is designed to allow effective high-reSOLUTIONon polishing of MAb in the second or third step of a purification scheme downstream of the protein A capture step.