Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScreen MabSelectTM Prism
Catalog No. 502908090
Encompass
This protein A column is prepacked with MabSelect PrismA chromatography resin. This affinity resin has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein ligand allowing efficient cleaning between runs. This allows future demands in monoclonal antibody purification to be met including processing of many bispecific antibodies.Enhanced dynamic binding capacity compared with other protein A chromatography resins.Excellent alkaline stability enables efficient cleaning and sanitization using 0.5-1.0 M NaOH.Convenient method optimization and parameter screening enabled by the 10 cm bed height for efficient chromatography process development.The enhanced alkaline stability of MabSelect PrismA also provides opportunities for more robust mAb purification using periodic counter-current chromatography. The possibility for cleaning using 1 M NaOH increases bioburden control for continuous processes which can be exposed to days or we
Catalog No. 50-290-8090 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17549815
This protein A column is prepacked with MabSelect PrismA chromatography resin. This affinity resin has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein ligand allowing efficient cleaning between runs. This allows future demands in monoclonal antibody purification to be met including processing of many bispecific antibodies.Enhanced dynamic binding capacity compared with other protein A chromatography resins.Excellent alkaline stability enables efficient cleaning and sanitization using 0.5-1.0 M NaOH.Convenient method optimization and parameter screening enabled by the 10 cm bed height for efficient chromatography process development.The enhanced alkaline stability of MabSelect PrismA also provides opportunities for more robust mAb purification using periodic counter-current chromatography. The possibility for cleaning using 1 M NaOH increases bioburden control for continuous processes which can be exposed to days or we