Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScreenTM CaptoTM Q ImpRe
Catalog No. 502901680
Encompass
HiScreen(TM) Capto(TM) Q ImpRes columns are prepacked with Capto(TM) Q ImpRes ion exchange chromatography medium and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.
Catalog No. 50-290-1680 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17547015
HiScreen(TM) Capto(TM) Q ImpRes columns are prepacked with Capto(TM) Q ImpRes ion exchange chromatography medium and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.