Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScreenTM MabSelect SuReT
Catalog No. 502908013
Encompass
HiScreen(TM) MabSelect(TM) SuRe(TM) LX is prepacked with MabSelect(TM) SuRe(TM) LX and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies when a high binding capacity is needed.
Catalog No. 50-290-8013 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17547415
HiScreen(TM) MabSelect(TM) SuRe(TM) LX is prepacked with MabSelect(TM) SuRe(TM) LX and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies when a high binding capacity is needed.