Learn More
Sigma Aldrich Fine Chemicals Biosciences HiScreenTM MabSelectTM SuR
Catalog No. 502907370
Encompass
HiScreen(TM) MabSelect(TM) SuRe(TM) is prepacked with MabSelect(TM) SuRe(TM) and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies (MAbs).
Catalog No. 50-290-7370 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28926977
HiScreen(TM) MabSelect(TM) SuRe(TM) is prepacked with MabSelect(TM) SuRe(TM) and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies (MAbs).