Learn More
VAZYME INTERNATIONAL LLC All-in-one RT Mix
Catalog No. NC3709631
Encompass
HiScript IV All-in-One Ultra RT SuperMix for qPCR is a next-generation qPCR-specific reverse transcription kit. This product integrates all the necessary components for genomic DNA removal and RNA reverse transcription in a single tube, requiring only the addition of template and water to initiate the reaction. With specially optimized reverse transcriptase, DNase, and a proprietary buffer, it allows for rapid completion of both genomic DNA removal and reverse transcription in just 5 minutes. The resulting cDNA is compatible with both dye-based and probe-based qPCR methods.
Catalog No. NC3709631 Supplier VAZYME INTERNATIONAL LLC Supplier No. R43301
HiScript IV All-in-One Ultra RT SuperMix for qPCR is a next-generation qPCR-specific reverse transcription kit. This product integrates all the necessary components for genomic DNA removal and RNA reverse transcription in a single tube, requiring only the addition of template and water to initiate the reaction. With specially optimized reverse transcriptase, DNase, and a proprietary buffer, it allows for rapid completion of both genomic DNA removal and reverse transcription in just 5 minutes. The resulting cDNA is compatible with both dye-based and probe-based qPCR methods.