Learn More
CUBE BIOTECH HisCube Ni-INDIGO MagBead MINI Kit; 1 kit
Catalog No. NC2684677
Encompass
Ready to use kit for His-tag protein purification kit, for 50x purifications. 1x10 ml INDIGO Agarose, 2x50ml Binding Buffer, 2x50ml Washing Buffer,1 MagBead separator, 50 tubes, 1 x 500mM DTT, 1x500mM EDTA + Protocols
Catalog No. NC2684677 Supplier CUBE BIOTECH Supplier No. 80201
Ready to use kit for His-tag protein purification kit, for 50x purifications. 1x10 ml INDIGO Agarose, 2x50ml Binding Buffer, 2x50ml Washing Buffer,1 MagBead separator, 50 tubes, 1 x 500mM DTT, 1x500mM EDTA + Protocols