Learn More
Seracare Life Sciences Inc HISDETECTOR WB KIT AP COLOR
Supplier:  Seracare Life Sciences Inc 58100025
Encompass_Preferred
HisDetector Western Blot Kit, AP Colorimetric uses Nickel-AP and BCIP/NBT 1-component substrate to produce a deep purple band at the site of reaction. Kit utilizes the specificity and high affinity of Nickel Conjugates (read more about Nickel conjugates) and optimized reagents to provide picogram level sensitivity with low background.
Catalog No. 50-673-81