Learn More
Seracare Life Sciences Inc HISDETECTOR WB KIT HRP CHEMI
Catalog No. 5067379
Encompass_Preferred
HisDetector Western Blot Kit, HRP Chemiluminescent uses Nickel-HRP and LumiGLO HRP Chemiluminescent Substrate for rapid, sensitive detection of His-tagged proteins. Kit utilizes the specificity and high affinity of Nickel Conjugates and optimized reagents to provide picogram level sensitivity with low background. See our Application Note for more information on the performance of HisDetector Western Blot Kits. Sufficient reagents provided for running 40 blots (10 x 10 cm2 blots).
This item is not returnable. View return policy
Catalog No. 50-673-79 Supplier Seracare Life Sciences Inc Supplier No. 58100024
HisDetector Western Blot Kit, HRP Chemiluminescent uses Nickel-HRP and LumiGLO HRP Chemiluminescent Substrate for rapid, sensitive detection of His-tagged proteins. Kit utilizes the specificity and high affinity of Nickel Conjugates and optimized reagents to provide picogram level sensitivity with low background. See our Application Note for more information on the performance of HisDetector Western Blot Kits. Sufficient reagents provided for running 40 blots (10 x 10 cm2 blots).