Learn More
Serim Research Corp Serim GUARDIAN™ HiSENSE ULTRA 0.1 Test Strips
SDP
Catalog No. NC1039969
Encompass_Preferred
Serim  HiSENSE ULTRA 0.1™ Test Strips provide a quick and convenient semi-quantitative test for indicating low levels of total chlorine(chloramines/free chlorine) in feed water used to prepare dialysate. Kit contains 5 bottles of 100 tests plus 5 sample cups
Catalog No. NC1039969 Supplier Serim Research Corp Supplier No. 5167
Serim  HiSENSE ULTRA 0.1™ Test Strips provide a quick and convenient semi-quantitative test for indicating low levels of total chlorine(chloramines/free chlorine) in feed water used to prepare dialysate. Kit contains 5 bottles of 100 tests plus 5 sample cups