Learn More
Cayman Chemical His-Tag Detection ELISA Kit, 96 strip wells
SDP
Supplier:  Cayman Chemical 1001244596STRIPWELLS
Encompass_Preferred
Cayman’s His-Tag Detection ELISA is competitive assay designed for the rapid, semi-quantitative screening of cell lysates and affinity column fractions for His-tagged proteins. It is intended to serve as a substitute for SDS-PAGE, thereby expediting the screening of affinity column fractions.
Catalog No. NC1368322