Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3005MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 1415-1. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma. Cellular Staining: Nuclear. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Nuclei of human leukemia biopsy cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~30kDa.Chromosome: 3q21-q22; 6q21-q22; 22q13.1. Gene Symbol: H1. Gene ID: 3005. Ensemble: ENSG00000189060. Uniprot: NP_005309.
Catalog No. 50-188-4765