Learn More
Biomatik Corporation HIS (Histamine) ELISA Kit
Supplier:  Biomatik Corporation EKF58487
Encompass_Preferred
HIS (Histamine) ELISA Kit for detecting HIS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Histamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7406