Learn More
Medchemexpress LLC HY-102040 5mg Medchemexpress, Hispidol CAS:5786-54-9 Purity:>98%
SDP
Catalog No. 501967967
Encompass_Preferred
Medchemexpress, HY-102040 5mg Hispidol CAS:5786-54-9 Hispidol ((Z)-Hispidol) is a potential therapeutic for inflammatory bowel disease; inhibits TNF-α induced adhesion of monocytes to colon epithelial cells with an IC50 of 0.50 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7967 Supplier Medchemexpress LLC Supplier No. HY1020405MG
Medchemexpress, HY-102040 5mg Hispidol CAS:5786-54-9 Hispidol ((Z)-Hispidol) is a potential therapeutic for inflammatory bowel disease; inhibits TNF-α induced adhesion of monocytes to colon epithelial cells with an IC50 of 0.50 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.