Learn More
Enzo Life Sciences Hispolon (25mg). CAS: 173933-40-9
SDP
Supplier:  Enzo Life Sciences ALX350384M025
Encompass_Preferred
Polyphenolic compound. Induces ROS-mediated apoptosis. Has antiviral activity. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8876