Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Spin Columns, 3 mL

Catalog No. PI88226 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88226 5 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88226 Supplier Thermo Scientific™ Supplier No. 88226
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin

  • Versatile – purify proteins using native or denaturing conditions

  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives

  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges

  • Cost-effective – reuse the same batch of resin at least five times

  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns

TRUSTED_SUSTAINABILITY

Specifications

Description HisPur Ni-NTA Spin Columns, 3mL
Content And Storage Store at 4°C
Form Liquid Suspension
Product Line HisPur
Quantity 5 Columns
Target His-Tagged Protein
Type Spin Column
Stationary Phase Divalent Nickel, Metal Chelate
Format Spin Column
Column Type Affinity, Agarose Resin
Capacity (Metric) 3 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.