Learn More
Epigentek Inc Histone H3K9me2 (H3K9 Dimethyl) Polyclonal Antibody (25 ul)
SDP
Supplier:  Epigentek Inc A4035025
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human DiMethyl-Histone H3-K9. Tested Applications: WB, IHC, IF, IP, ChIP, ChIPseq. Reactivity: Broad Range, Mouse, Rat, Human. Alternative Names: HIST3H3, H3/g.
Catalog No. 50-109-5512