Learn More
Epigentek Inc Histone H3K9me3 (H3K9 Trimethyl) Monoclonal Antibody [2C3] (50 ul)
SDP
Supplier:  Epigentek Inc A68391050
Encompass_Preferred
Unconjugated Monoclonal Antibody Raised in Mouse. Immunogen: Synthetic Peptide. Tested Applications: ELISA, WB. Reactivity: Human, Mouse, Rat. Alternative Names: HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; Histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/.
Catalog No. 50-125-0787