Learn More
Epigentek Inc Histone H4R3 Dimethyl Asymmetric (H4R3me2a) Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A3708100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human Asymmetric DiMethyl-Histone H3-R17. Tested Applications: WB, IHC, IF, IP. Reactivity: Human, Mouse, Rat, Broad Range. Alternative Names: H3R17me2a, HIST1H3J, H3/j , H3FJ , Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h , Histone H3/I, Histone H3/j , Histone H3/k, Histone H3/l, H3 Arginine 17 me2a.
Catalog No. 50-109-5398