Learn More
Aviva Systems Biology Corp HIST1H1C Antibody - middle region (ARP46122_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46122_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST1H1C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIST1H1C. Alternate names: H1.2, H1F2, MGC3992, H1CApplicationCHIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53460