Learn More
Aviva Systems Biology Corp HIST1H2AE Antibody - middle region (ARP60986_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60986_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST1H2AE. It was validated on Western Blot. . Alternate names: H2A.1, H2A.2, H2A/a, H2AFA, HIST1H2ABApplicationWB. Species reactivity: Cow, Human, Pig, Rat
Catalog No. 50-287-225