Learn More
Aviva Systems Biology Corp HIST1H2AH Antibody - C-terminal region (ARP60873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST1H2AH. It was validated on Western Blot. . Alternate names: H2A/S, H2AFALii, MGC171151, dJ86C11.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-195