Learn More
Aviva Systems Biology Corp HIST1H2BK ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP60871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST1H2BK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HIST1H2BK. Alternate names: H2B/S, H2BFAiii, H2BFT, MGC131989ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Mouse, Rabbit, Rat
Catalog No. 50-287-193