Learn More
Aviva Systems Biology Corp HIST1H3A Antibody - N-terminal region (ARP60982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST1H3A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HIST1H3A. Alternate names: H3/A, H3FA, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3JApplicationWB. Species reactivity: Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-287-222