Learn More
Apexbio Technology LLC Histamine 2HCl 56-92-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092639
Encompass_Preferred
Histamine 2HCl (56-92-8) is a small-molecule agonist targeting histamine H1 and H2 receptors It is designed to selectively stimulate these receptors thereby modulating downstream intracellular signaling pathways Histamine 2HCl exerts its biological activity primarily through receptor agonism triggering pathways involved in neurotransmission gastric acid secretion vascular permeability and inflammation In receptor binding assays and cell-based experiments Histamine 2HCl demonstrates receptor stimulation with reported IC50 values for H1 and H2 receptor activation typically ranging from micromolar to sub-micromolar concentrations Based on these pharmacological properties Histamine 2HCl holds research potential in studies of allergic reactions gastrointestinal physiology and central nervous system functions
Catalog No. 50-000-92639 Supplier Apexbio Technology LLC Supplier No. B15615.1
Histamine 2HCl (56-92-8) is a small-molecule agonist targeting histamine H1 and H2 receptors It is designed to selectively stimulate these receptors thereby modulating downstream intracellular signaling pathways Histamine 2HCl exerts its biological activity primarily through receptor agonism triggering pathways involved in neurotransmission gastric acid secretion vascular permeability and inflammation In receptor binding assays and cell-based experiments Histamine 2HCl demonstrates receptor stimulation with reported IC50 values for H1 and H2 receptor activation typically ranging from micromolar to sub-micromolar concentrations Based on these pharmacological properties Histamine 2HCl holds research potential in studies of allergic reactions gastrointestinal physiology and central nervous system functions