Learn More
Medchemexpress LLC Histamine | 51-45-6 | 99.0% | 10MM/1ML
SDP
Catalog No. 5000413600
Encompass_Preferred
Histamine is an organic nitrogen compound that acts as an agonist for histamine receptors and a vasodilator. It participates in local immune responses, regulates intestinal physiological functions, and functions as a neurotransmitter. It influences the p38 MAPK/Akt signaling pathway and exhibits antitumor, antioxidant, and anti-inflammatory properties. It can be utilized in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.
  • Acts as an agonist for histamine receptor and a vasodilator.
  • Involved in local immune responses, regulation of intestinal physiological functions, and neurotransmission.
  • Affects the p38 MAPK/Akt signaling pathway.
  • Demonstrates antitumor, antioxidant, and anti-inflammatory activities.
  • Applicable in research related to acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.
Catalog No. 50-004-13600 Supplier Medchemexpress LLC Supplier No. HYB120410MM/1ML
Histamine is an organic nitrogen compound that acts as an agonist for histamine receptors and a vasodilator. It participates in local immune responses, regulates intestinal physiological functions, and functions as a neurotransmitter. It influences the p38 MAPK/Akt signaling pathway and exhibits antitumor, antioxidant, and anti-inflammatory properties. It can be utilized in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.
  • Acts as an agonist for histamine receptor and a vasodilator.
  • Involved in local immune responses, regulation of intestinal physiological functions, and neurotransmission.
  • Affects the p38 MAPK/Akt signaling pathway.
  • Demonstrates antitumor, antioxidant, and anti-inflammatory activities.
  • Applicable in research related to acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.